Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166388.1 | NP_001159860.1 | 2 | 1528 | ||
NM_001166389.1 | NP_001159861.1 | 2 | 1259 | |||
NM_008975.3 | NP_033001.2 | 2 | 1328 | |||
GenBank mRNA | AF035645.1 | - | 1 | 309 | ||
AK003954.1 | - | 1 | 66 | |||
AK004562.1 | - | 2 | 938 | |||
AK014601.1 | - | 2 | 942 | |||
AK143702.1 | - | 2 | 980 | |||
AK147489.1 | - | 1 | 3992 | |||
AK172192.1 | - | 2 | 982 | |||
BC027445.1 | - | 1 | 139 | 4482106 | ||
BC066043.1 | - | 2 | 1528 | 6415021 | ||
CT010215.1 | - | 1 | 94 |