Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011201.3 | NP_035331.3 | 6, 7 | 867 | ||
GenBank mRNA | AK149979.1 | - | 6, 7 | 823 | ||
AK150322.1 | - | 6, 7 | 823 | |||
AK151371.1 | - | 6, 7 | 780 | |||
AK153088.1 | - | 6, 7 | 805 | |||
AK156281.1 | - | 6, 7 | 829 | |||
AK157431.1 | - | 6, 7 | 868 | |||
AK165567.1 | - | 6, 7 | 832 | |||
AK165949.1 | - | 7, 8 | 947 | |||
AK166020.1 | - | 6, 7 | 823 | |||
AK168255.1 | - | 6, 7 | 805 | |||
AK168772.1 | - | 6, 7 | 822 | |||
AK170620.1 | - | 6, 7 | 823 | |||
AK171380.1 | - | 6, 7 | 823 | |||
AK172206.1 | - | 7, 8 | 940 | |||
AK185819.1 | - | 1, 2 | 50 | |||
AK208930.1 | - | 1, 2 | 50 | |||
BC005729.1 | - | 1, 2 | 220 | |||
BC010191.1 | - | 6, 7 | 785 | 3156854 | ||
M97590.1 | - | 6, 7 | 783 |