Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 7 | 1145 | ||
GenBank mRNA | AF170902.1 | - | 6 | 712 | ||
AF170903.1 | - | 6 | 712 | |||
AK028783.1 | - | 7 | 1109 | |||
AK037646.1 | - | 7 | 1146 | |||
AK086524.1 | - | 7 | 1139 | |||
AK156480.1 | - | 7 | 891 | |||
D31842.1 | - | 6 | 742 |