Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111316.2 | NP_001104786.2 | 9 | 917 | ||
NM_001268286.1 | NP_001255215.1 | 6 | 526 | |||
NM_011210.4 | NP_035340.3 | 7 | 598 | |||
GenBank mRNA | AK054056.1 | - | 7 | 599 | ||
AK088875.1 | - | 1 | 3182 | |||
AK154893.1 | - | 6 | 521 | |||
BC028512.1 | - | 7 | 538 | |||
BC131655.1 | - | 7 | 538 | |||
L36091.1 | - | 2 | 124 | |||
M14342.1 | - | 7 | 560 | |||
M14343.1 | - | 3 | 1091 | |||
M92933.1 | - | 9 | 917 |