Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111316.2 | NP_001104786.2 | 13 | 1498 | ||
NM_001268286.1 | NP_001255215.1 | 10 | 1107 | |||
NM_011210.4 | NP_035340.3 | 11 | 1179 | |||
GenBank mRNA | AK054056.1 | - | 11 | 1180 | ||
AK154893.1 | - | 10 | 1102 | |||
BC131655.1 | - | 11 | 1119 | |||
L36091.1 | - | 6 | 705 | |||
M14342.1 | - | 11 | 1141 | |||
M14343.1 | - | 7 | 1672 | |||
M92933.1 | - | 13 | 1498 |