Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016933.3 | NP_058629.2 | 2 | 398 | ||
GenBank mRNA | AA265746.1 | - | 2 | 397 | ||
AI323196.1 | - | 2 | 370 | |||
AI627045.1 | - | 1 | 33 | |||
AK088334.1 | - | 2 | 396 | |||
AK165203.1 | - | 2 | 396 | |||
BC013273.1 | - | 2 | 397 | 4208992 | ||
BC100560.1 | - | 2 | 379 | 832557 | ||
D10105.1 | - | 2 | 374 | |||
U03856.1 | - | 1 | 391 |