Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011213.2 | NP_035343.2 | 20 | 4131 | ||
GenBank mRNA | AF300943.1 | - | 18 | 3794 | ||
BC057166.1 | - | 12 | 2682 | |||
CA315396.1 | - | 2 | 347 |