Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135657.1 | NP_001129129.1 | 10 | 2153 | ||
NM_008982.5 | NP_033008.3 | 10 | 2437 | |||
GenBank mRNA | AK147318.1 | - | 10 | 2358 | ||
AK147556.1 | - | 10 | 2358 | |||
AY038891.1 | - | 10 | 2255 | |||
AY039232.1 | - | 10 | 2255 | |||
D45212.1 | - | 10 | 2393 | |||
D83203.1 | - | 10 | 2154 | |||
DQ133576.1 | - | 10 | 2168 |