Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011215.2 | NP_035345.2 | 19 | 2770 | ||
GenBank mRNA | AK138655.1 | - | 19 | 2762 | ||
BC133678.1 | - | 19 | 2654 | 40109882 | ||
U57345.1 | - | 19 | 2657 | |||
U82439.1 | - | 14 | 1778 |