Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011215.2 | NP_035345.2 | 9 | 1373 | ||
GenBank mRNA | AK081267.1 | - | 9 | 1366 | ||
AK138655.1 | - | 9 | 1365 | |||
AK220255.1 | - | 9 | 1301 | |||
BC133678.1 | - | 9 | 1257 | 40109882 | ||
U57345.1 | - | 9 | 1260 |