Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164401.1 | NP_001157873.1 | 22 | 3577 | ||
NM_001164402.1 | NP_001157874.1 | 12 | 1207 | |||
NM_001164403.1 | NP_001157875.1 | 11 | 1123 | |||
NM_011216.3 | NP_035346.3 | 23 | 3661 | |||
GenBank mRNA | AF135166.1 | - | 22 | 3542 | ||
AF295638.1 | - | 23 | 3274 | |||
BC051976.1 | - | 12 | 1120 | |||
BC052743.1 | - | 23 | 3495 | 6834312 | ||
BC063258.1 | - | 13 | 1316 | |||
BC079649.1 | - | 1123 | ||||
U37465.1 | - | 12 | 1191 | |||
U37466.1 | - | 11 | 1107 |