Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164401.1 | NP_001157873.1 | 12 | 2511 | ||
NM_011216.3 | NP_035346.3 | 12 | 2511 | |||
GenBank mRNA | AF135166.1 | - | 12 | 2476 | ||
AF295638.1 | - | 12 | 2124 | |||
BC052743.1 | - | 12 | 2345 | 6834312 | ||
BC063258.1 | - | 2 | 166 |