Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008991.2 | NP_033017.2 | 9 | 892 | ||
GenBank mRNA | AK004660.1 | - | 9 | 873 | ||
AK144060.1 | - | 9 | 874 | |||
AK153298.1 | - | 9 | 850 | |||
BC009119.1 | - | 9 | 884 | 3154529 | ||
BC054446.1 | - | 9 | 840 | 6477711 | ||
CB245413.1 | - | 4 | 340 |