Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278447.1 | NP_001265376.1 | 6 | 530 | ||
NM_181070.6 | NP_851415.1 | 6 | 512 | |||
GenBank mRNA | AB232612.1 | - | 6 | 414 | ||
AK028155.1 | - | 2 | 113 | |||
AK045514.1 | - | 6 | 475 | |||
AK140901.1 | - | 6 | 480 | |||
AK143673.1 | - | 6 | 488 | |||
AK146177.1 | - | 6 | 437 | |||
AK146397.1 | - | 6 | 437 | |||
AK154920.1 | - | 6 | 489 | |||
AK216482.1 | - | 3 | 224 | |||
BC056351.1 | - | 6 | 466 | 6807284 | ||
L04966.1 | - | 6 | 418 | |||
X80333.1 | - | 6 | 414 |