Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011227.1 | NP_035357.1 | 2 | 644 | ||
GenBank mRNA | AB232614.1 | - | 2 | 644 | ||
AK145566.1 | - | 2 | 821 | |||
AK152454.1 | - | 2 | 770 | |||
AK163743.1 | - | 2 | 784 | |||
AK164541.1 | - | 2 | 784 | |||
BC089570.1 | - | 2 | 769 | 6747570 | ||
X80332.1 | - | 2 | 644 |