Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016858.2 | NP_058554.1 | 2 | 885 | ||
GenBank mRNA | AB004665.1 | - | 2 | 835 | ||
AB232629.1 | - | 2 | 519 | |||
AK004974.1 | - | 2 | 621 | |||
AK087967.1 | - | 2 | 625 | |||
AK139821.1 | - | 2 | 871 | |||
AK144706.1 | - | 2 | 624 | |||
AK159674.1 | - | 2 | 620 | |||
AK163783.1 | - | 2 | 872 | |||
AK170483.1 | - | 2 | 514 | |||
BC065076.1 | - | 2 | 885 | 6815992 |