Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166399.2 | NP_001159871.1 | 2 | 269 | ||
NM_009001.6 | NP_033027.1 | 2 | 264 | |||
GenBank mRNA | AB232587.1 | - | 1 | 79 | ||
AK005307.1 | - | 2 | 226 | |||
AK005362.1 | - | 2 | 226 | |||
AK158727.1 | - | 1 | 424 | |||
AK161975.1 | - | 2 | 233 | |||
BC018451.1 | - | 1 | 472 | 3489400 | ||
BC053519.1 | - | 2 | 228 | 6467763 | ||
BY097011.1 | - | 2 | 265 | |||
BY251171.1 | - | 2 | 258 | |||
BY258862.1 | - | 2 | 264 |