Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166399.2 | NP_001159871.1 | 3 | 512 | ||
NM_009001.6 | NP_033027.1 | 3 | 507 | |||
GenBank mRNA | AB232587.1 | - | 2 | 322 | ||
AK005307.1 | - | 3 | 469 | |||
AK005362.1 | - | 3 | 469 | |||
AK158727.1 | - | 2 | 667 | |||
AK161975.1 | - | 3 | 476 | |||
BC018451.1 | - | 1 | 1077 | 3489400 | ||
BC053519.1 | - | 3 | 471 | 6467763 |