Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB232595.1 | - | 4 | 510 | ||
AK089199.1 | - | 5 | 766 | |||
AK134691.1 | - | 5 | 651 | |||
AK159832.1 | - | 5 | 701 | |||
AK167599.1 | - | 5 | 701 | |||
AK170540.1 | - | 5 | 721 | |||
AK172328.1 | - | 5 | 658 | |||
BC023027.1 | - | 5 | 667 | 5369566 | ||
BC027378.1 | - | 4 | 473 | |||
BC029678.1 | - | 5 | 651 | 5368061 |