Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163663.1 | NP_001157135.1 | 5 | 1009 | ||
NM_024287.4 | NP_077249.1 | 5 | 1009 | |||
GenBank mRNA | AB041575.1 | - | 5 | 808 | ||
AB232596.1 | - | 5 | 371 | |||
AK051246.1 | - | 5 | 844 | |||
AK083262.1 | - | 5 | 833 | |||
AK084131.1 | - | 5 | 817 | |||
AK154012.1 | - | 5 | 906 | |||
AK160134.1 | - | 5 | 888 | |||
AK160866.1 | - | 5 | 817 | |||
AK169842.1 | - | 5 | 1010 | |||
AK183477.1 | - | 4 | 292 | |||
AK185691.1 | - | 4 | 292 | |||
AK197621.1 | - | 11 | ||||
BC019118.1 | - | 5 | 641 | 3711087 | ||
BC026915.1 | - | 4 | 299 |