Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293652.1 | NP_001280581.1 | 5 | 564 | ||
NM_001293653.1 | NP_001280582.1 | 3 | 225 | |||
NM_001293654.1 | NP_001280583.1 | 4 | 391 | |||
NM_001293655.1 | NP_001280584.1 | 3 | 304 | |||
NM_009005.3 | NP_033031.2 | 3 | 342 | |||
GenBank mRNA | AB232599.1 | - | 2 | 136 | ||
AK005004.1 | - | 3 | 304 | |||
AK057671.1 | - | 3 | 271 | |||
AK135659.1 | - | 4 | 392 | |||
AK139914.1 | - | 3 | 295 | |||
AK148117.1 | - | 3 | 335 | |||
AK151078.1 | - | 3 | 316 | |||
AK152666.1 | - | 5 | 539 | |||
AK154234.1 | - | 3 | 323 | |||
AK158971.1 | - | 3 | 226 | |||
AK159507.1 | - | 3 | 272 | |||
AK159928.1 | - | 3 | 243 | |||
BC004597.1 | - | 3 | 260 | 3584891 | ||
BC086793.1 | - | 3 | 258 | 6414012 | ||
BY291181.1 | - | 3 | 343 | |||
CT010352.1 | - | 2 | 136 | |||
X89650.1 | - | 3 | 292 |