Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163478.1 | NP_001156950.1 | 8 | 824 | ||
NM_001163479.1 | NP_001156951.1 | 6 | 762 | |||
NM_011231.2 | NP_035361.2 | 7 | 962 | |||
GenBank mRNA | AF231926.1 | - | 8 | 900 | ||
AK010690.1 | - | 6 | 514 | |||
AK159902.1 | - | 9 | 1270 | |||
AK160162.1 | - | 8 | 811 | |||
BC003948.1 | - | 8 | 887 | |||
BC021335.1 | - | 7 | 808 | |||
BC057661.1 | - | 7 | 1476 | |||
BC132473.1 | - | 7 | 840 | 40130750 | ||
BC138547.1 | - | 7 | 840 | 8861567 | ||
BC145323.1 | - | 8 | 922 | |||
BC145324.1 | - | 7 | 824 | |||
BC145325.1 | - | 8 | 938 | |||
BC145326.1 | - | 8 | 898 | |||
CN456589.1 | - | 6 | 671 | |||
U12922.1 | - | 7 | 818 |