Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163478.1 | NP_001156950.1 | 6, 7 | 617 | ||
NM_001163479.1 | NP_001156951.1 | 4, 5 | 555 | |||
NM_011231.2 | NP_035361.2 | 5, 6 | 755 | |||
GenBank mRNA | AK010690.1 | - | 4, 5 | 307 | ||
AK159902.1 | - | 7, 8 | 1063 | |||
AK160162.1 | - | 6, 7 | 604 | |||
BC021335.1 | - | 5, 6 | 601 | |||
BC132473.1 | - | 5, 6 | 633 | 40130750 | ||
BC138547.1 | - | 5, 6 | 633 | 8861567 | ||
BC145325.1 | - | 6, 7 | 731 | |||
BC145326.1 | - | 6, 7 | 691 | |||
CN456589.1 | - | 4, 5 | 464 | |||
U12922.1 | - | 5, 6 | 611 |