Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009009.4 | NP_033035.3 | 14 | 2110 | ||
GenBank mRNA | AF332085.1 | - | 13 | 1865 | ||
AF332086.1 | - | 13 | 1865 | |||
AK004746.1 | - | 14 | 1913 | |||
AK128984.1 | - | 14 | 1999 | |||
AK129050.1 | - | 9 | 1238 | |||
AK139492.1 | - | 13 | 1763 | |||
AK145591.1 | - | 14 | 2077 | |||
AK163992.1 | - | 1971 | ||||
AK168718.1 | - | 14 | 2098 | |||
AK168896.1 | - | 14 | 2059 | |||
BC043032.1 | - | 13 | 1879 | 5696540 | ||
BC129916.1 | - | 13 | 1909 | 40111820 | ||
BC129917.1 | - | 13 | 1910 | 40111821 | ||
D37790.1 | - | 13 | 1798 | |||
X98293.1 | - | 13 | 1865 |