Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011234.4 | NP_035364.1 | 4 | 460 | ||
GenBank mRNA | AK011242.1 | - | 4 | 461 | ||
AK045050.1 | - | 4 | 463 | |||
AK076468.1 | - | 4 | 408 | |||
AK148165.1 | - | 460 | ||||
AK151157.1 | - | 4 | 439 | |||
AK151177.1 | - | 4 | 439 | |||
AK160282.1 | - | 4 | 452 | |||
BC027384.1 | - | 4 | 425 | 4951015 | ||
D13473.1 | - | 4 | 415 | |||
D13803.1 | - | 4 | 428 |