Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009013.3 | NP_033039.2 | 3 | 150 | ||
GenBank mRNA | AK011379.1 | - | 3 | 151 | ||
AK077829.1 | - | 3 | 183 | |||
AK079525.1 | - | 3 | 172 | |||
AK166703.1 | - | 3 | 183 | |||
BC003738.1 | - | 3 | 164 | 3495632 |