Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128148.2 | NP_001121620.1 | 9 | 1124 | ||
NM_001313965.1 | NP_001300894.1 | 8 | 922 | |||
NM_001313966.1 | NP_001300895.1 | 7 | 726 | |||
NM_003234.3 | NP_003225.2 | 9 | 1265 | |||
GenBank mRNA | AB209254.1 | - | 9 | 1028 | ||
AK291723.1 | - | 9 | 1265 | |||
AK294758.1 | - | 7 | 656 | |||
BC001188.1 | - | 9 | 1038 | 3354176 | ||
BX537966.1 | - | 8 | 852 | |||
M11507.1 | - | 9 | 1077 | |||
X01060.1 | - | 9 | 1245 |