Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 10, 11 | 966 | ||
NM_033475.3 | NP_258436.2 | 9, 10 | 1311 | |||
GenBank mRNA | AB082927.1 | - | 9, 10 | 1224 | ||
AB232630.1 | - | 9, 10 | 722 | |||
AF322068.1 | - | 9, 10 | 722 | |||
AF327929.1 | - | 10, 11 | 916 | |||
AK160308.1 | - | 1280 | ||||
AK181473.1 | - | 2, 3 | 136 | |||
BC038638.1 | - | 9, 10 | 786 | 5058535 | ||
S72304.1 | - | 7, 8 | 569 |