Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009022.4 | NP_033048.2 | 3, 4 | 416 | ||
GenBank mRNA | AK078553.1 | - | 3, 4 | 357 | ||
BC075704.1 | - | 3, 4 | 383 | 30471325 | ||
CJ166631.1 | - | 3, 4 | 417 | |||
X99273.1 | - | 3, 4 | 361 |