Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139511.1 | NP_001132983.1 | 4 | 876 | ||
NM_001139512.1 | NP_001132984.1 | 5 | 642 | |||
NM_001139513.1 | NP_001132985.1 | 5 | 924 | |||
NM_023130.3 | NP_075619.2 | 5 | 959 | |||
GenBank mRNA | AK014356.1 | - | 5 | 712 | ||
AK135386.1 | - | 4 | 635 | |||
AK152290.1 | - | 5 | 718 | |||
AK154787.1 | - | 4 | 633 | |||
AK168781.1 | - | 5 | 716 | |||
BC004851.1 | - | 4 | 620 | 2810841 | ||
BC016587.1 | - | 5 | 708 | 2811315 | ||
L17076.1 | - | 5 | 689 | |||
S72641.1 | - | 5 | 910 |