Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011240.3 | NP_035370.2 | 9, 10 | 1393 | ||
GenBank mRNA | AK145306.1 | - | 9, 10 | 1324 |