Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009023.3 | NP_033049.2 | 6 | 1117 | ||
GenBank mRNA | AK009672.1 | - | 5 | 961 | ||
AK213318.1 | - | 2 | 53 | |||
BC111863.1 | - | 6 | 1073 | 40046186 | ||
J03962.1 | - | 6 | 1053 | |||
X15788.1 | - | 6 | 1106 |