Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001176528.1 | NP_001169999.1 | 3 | 847 | ||
NM_001177302.1 | NP_001170773.1 | 5 | 1052 | |||
NM_001177303.1 | NP_001170774.1 | 4 | 797 | |||
NM_009024.2 | NP_033050.2 | 4 | 919 | |||
GenBank mRNA | AB221572.1 | - | 3 | 366 | ||
AK042163.1 | - | 4 | 798 | |||
AK153644.1 | - | 3 | 393 | |||
AK154623.1 | - | 4 | 892 | |||
AK170956.1 | - | 3 | 847 | |||
BC010216.1 | - | 4 | 917 | 3498142 | ||
BC038266.1 | - | 3 | 916 | |||
BC040383.1 | - | 5 | 1052 | |||
BC090396.1 | - | 3 | 463 | |||
CT010231.1 | - | 3 | 362 | |||
S56656.1 | - | 4 | 899 |