Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009027.3 | NP_033053.2 | 968 | |||
GenBank mRNA | AK041764.1 | - | 311 | |||
AK158472.1 | - | 1133 | ||||
U67326.2 | - | 968 |