Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013833.2 | NP_038861.2 | 1 | 409 | ||
GenBank mRNA | AF001906.1 | - | 1 | 411 | ||
AK053517.1 | - | 390 | ||||
AK133951.1 | - | 1 | 390 | |||
BC024731.1 | - | 1 | 409 | 5366450 | ||
BC058757.1 | - | 1 | 373 | 6492771 | ||
U73177.1 | - | 1 | 401 |