Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270516.1 | NP_001257445.1 | 9 | 1137 |
Gene Symbol: Gm10256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270510.1 | NP_001257439.1 | 1137 |
Gene Symbol: Gm10352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270511.1 | NP_001257440.1 | 1137 |
Gene Symbol: Gm21677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270513.1 | NP_001257442.1 | 1137 |
Gene Symbol: Gm21693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270514.1 | NP_001257443.1 | 1137 |
Gene Symbol: Gm21704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270515.1 | NP_001257444.1 | 1137 |
Gene Symbol: Gm3376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270512.1 | NP_001257441.1 | 9 | 1137 |
Gene Symbol: Gm4064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270518.1 | NP_001257447.1 | 9 | 1137 |
Gene Symbol: Rbmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166384.1 | NP_001159856.1 | 9 | 1137 | ||
NM_011253.2 | NP_035383.2 | 10 | 1288 | |||
GenBank mRNA | AK016258.1 | - | 1139 | |||
BC150670.1 | - | 1118 | 9087581 | |||
U36929.1 | - | 1129 | ||||
Y15131.1 | - | 10 | 1277 |