Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000577.4 | NP_000568.1 | 4 | 289 | ||
NM_001318914.1 | NP_001305843.1 | 6 | 523 | |||
NM_173841.2 | NP_776213.1 | 5 | 352 | |||
NM_173842.2 | NP_776214.1 | 3 | 281 | |||
NM_173843.2 | NP_776215.1 | 5 | 460 | |||
XM_005263661.4 | XP_005263718.1 | 4 | 308 | |||
XM_011511121.1 | XP_011509423.1 | 8 | 811 | |||
GenBank mRNA | AF043143.1 | - | 5 | 460 | ||
AK290898.1 | - | 5 | 342 | |||
AK290926.1 | - | 3 | 283 | |||
BC009745.2 | - | 4 | 194 | 3689284 | ||
BI766516.1 | - | 3 | 281 | |||
DA667440.1 | - | 6 | 440 | |||
M55646.1 | - | 4 | 285 | |||
X52015.1 | - | 3 | 240 | |||
X53296.1 | - | 3 | 225 | |||
X84348.1 | - | 5 | 258 |