Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015745.2 | NP_056560.1 | 1 | 1852 | ||
GenBank mRNA | BC017610.1 | - | 1 | 1852 | 4527589 | |
BC035273.1 | - | 1 | 284 | |||
BC040282.1 | - | 1 | 1836 | |||
BC042480.1 | - | 1 | 1814 | 5363924 | ||
BC083133.1 | - | 1 | 1871 | 6492729 |