Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159487.1 | NP_001152959.1 | 5 | 761 | ||
NM_011255.3 | NP_035385.1 | 5 | 448 | |||
GenBank mRNA | AK004839.1 | - | 5 | 446 | ||
AK008765.1 | - | 5 | 446 | |||
AK169338.1 | - | 4 | 295 | |||
BC031809.1 | - | 5 | 426 | 4193984 | ||
BC093529.1 | - | 5 | 688 | 5135440 | ||
M74527.1 | - | 4 | 376 | |||
U63146.1 | - | 4 | 376 |