Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009040.3 | NP_033066.1 | 4 | 1372 | ||
GenBank mRNA | AF030513.1 | - | 4 | 1372 | ||
AK149404.1 | - | 3 | 1116 | |||
AK164728.1 | - | 1 | 551 | |||
BC089612.1 | - | 4 | 1341 | 6763439 |