Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011259.1 | NP_035389.1 | 3 | 244 | ||
GenBank mRNA | AK008470.1 | - | 1 | 79 | ||
BC119153.1 | - | 3 | 244 | 8733902 | ||
D63356.1 | - | 3 | 244 | |||
D63357.1 | - | 3 | 193 |