Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011260.1 | NP_035390.1 | 5 | 430 | ||
GenBank mRNA | AK008446.1 | - | 5 | 426 | ||
AK008608.1 | - | 5 | 426 | |||
BC046602.1 | - | 1 | 1747 | |||
BC061139.1 | - | 5 | 428 | 30298640 | ||
D63361.1 | - | 5 | 430 |