Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020022.2 | NP_064406.1 | 9 | 913 | ||
GenBank mRNA | AK011353.1 | - | 9 | 829 | ||
AK011649.1 | - | 9 | 826 | |||
AK075997.1 | - | 9 | 825 | |||
AK154681.1 | - | 9 | 867 | |||
BC004812.1 | - | 6 | 585 | |||
BC023028.1 | - | 9 | 834 | 5374851 | ||
BC099370.1 | - | 9 | 877 | 30937690 |