Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020022.2 | NP_064406.1 | 4, 5 | 427 | ||
GenBank mRNA | AK011353.1 | - | 4, 5 | 343 | ||
AK011649.1 | - | 4, 5 | 341 | |||
AK075997.1 | - | 4, 5 | 339 | |||
AK154681.1 | - | 4, 5 | 381 | |||
BC004812.1 | - | 1, 2 | 99 | |||
BC023028.1 | - | 4, 5 | 348 | 5374851 | ||
BC099370.1 | - | 4, 5 | 391 | 30937690 |