Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166414.1 | NP_001159886.1 | 10 | 1200 | ||
NM_011265.3 | NP_035395.2 | 9 | 1241 | |||
GenBank mRNA | AK030008.1 | - | 11 | 1273 | ||
AK041120.1 | - | 9 | 1226 | |||
AK132267.1 | - | 5 | 566 | |||
AK140928.1 | - | 8 | 1050 | |||
BC017598.1 | - | 9 | 1215 | 4483833 |