Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009060.2 | NP_033086.1 | 5 | 649 | ||
GenBank mRNA | AK146465.1 | - | 5 | 681 | ||
BC012710.1 | - | 5 | 662 | 4210374 | ||
D86217.1 | - | 4 | 601 | |||
U28937.1 | - | 5 | 649 |