Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011271.2 | NP_035401.2 | 2 | 196 | ||
GenBank mRNA | AK075775.1 | - | 2 | 196 | ||
BC030036.1 | - | 2 | 176 | 1230895 | ||
M27814.1 | - | 2 | 163 |