Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286865.1 | NP_001273794.1 | 6, 7 | 733 | ||
NM_011275.3 | NP_035405.2 | 6, 7 | 733 | |||
GenBank mRNA | AF048993.1 | - | 6, 7 | 694 | ||
AK011680.1 | - | 5, 6 | 648 | |||
AK088032.1 | - | 4, 5 | 1248 | |||
BC019411.1 | - | 6, 7 | 662 | 5133942 | ||
BG084607.2 | - | 2, 3 | 183 | |||
BM943446.1 | - | 3, 4 | 253 |