Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286865.1 | NP_001273794.1 | 2, 3 | 330 | ||
NM_011275.3 | NP_035405.2 | 2, 3 | 330 | |||
GenBank mRNA | AF048993.1 | - | 2, 3 | 291 | ||
AK011680.1 | - | 2, 3 | 303 | |||
BC019411.1 | - | 2, 3 | 259 | 5133942 | ||
BY274268.1 | - | 2, 3 | 331 | |||
CA554498.1 | - | 2, 3 | 281 | |||
CJ073105.1 | - | 2, 3 | 306 |