Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019413.2 | NP_062286.2 | 2 | 1274 | ||
GenBank mRNA | AK032752.1 | - | 2 | 1240 | ||
AK138485.1 | - | 2 | 1274 | |||
AK142047.1 | - | 2 | 1220 | |||
AK143001.1 | - | 2 | 1274 | |||
Y17793.1 | - | 2 | 536 |